EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Poly(N-acryloylsarcosine methyl ester) protein-resistant surfaces.
PMID 16853712
Abstract
A new class of protein-resistant film based on N-substituted glycine derivatives is described. Pulsed plasma deposited poly(N-acryloylsarcosine methyl ester) coatings are shown to be resistant toward the adsorption of fibrinogen and lysozyme. Deposition and UV irradiation of the polymer through a masked grid are found to be effective ways for generating negative and positive image protein arrays, respectively, onto a range of different substrate materials.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
84570
Sarcosine methyl ester hydrochloride, ≥97.0% (T)
C4H9NO2 · HCl