EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Directed assembly and separation of self-assembled monolayers via electrochemical processing.
PMID 16854150
Abstract
Separated domains of 1-dodecanethiolate were fabricated via solution displacement of preformed 1-adamantanethiolate self-assembled monolayers on Au{111}. Subsequently, the 1-adamantanethiolate domains were desorbed selectively, and the substrate was exposed to a 1-octanethiol solution, creating artificially separated self-assembled monolayers of 1-dodecanethiolate and 1-octanethiolate. The molecular order of each lattice type and the apparent height differences imaged with scanning tunneling microscopy and the two distinct cathodic peaks observed with cyclic voltammetry indicated distinct separated domains of each lattice type in the separated self-assembled monolayers. By manipulating the intermolecular interaction strengths of the patterned molecules, we are able to control the structure and properties of the separated self-assembled monolayers via the exploitation of competitive adsorption and the utilization of electrochemical processing, which can be extended to other self-assembly patterning techniques such as microdisplacement printing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
471836
1-Octanethiol, ≥98.5%
C8H18S