EMAIL THIS PAGE TO A FRIEND
Applied spectroscopy
Multivariate curve resolution applied to infrared reflection measurements of soil contaminated with an organophosphorus analyte.
PMID 16854257
Abstract
Multivariate curve resolution (MCR) is a powerful technique for extracting chemical information from measured spectra of complex mixtures. A modified MCR technique that utilized both measured and second-derivative spectra to account for observed sample-to-sample variability attributable to changes in soil reflectivity was used to estimate the spectrum of dibutyl phosphate (DBP) adsorbed on two different soil types. This algorithm was applied directly to measurements of reflection spectra of soils coated with analyte without resorting to soil preparations such as grinding or dilution in potassium bromide. The results provided interpretable spectra that can be used to guide strategies for detection and classification of organic analytes adsorbed on soil. Comparisons to the neat DBP liquid spectrum showed that the recovered analyte spectra from both soils showed spectral features from methyl, methylene, hydroxyl, and P=O functional groups, but most conspicuous was the absence of the strong PO-(CH2)3CH3 stretch absorption at 1033 cm(-1). These results are consistent with those obtained previously using extended multiplicative scatter correction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68572
Dibutyl phosphate, ≥97.0% (T)
C8H19O4P