EMAIL THIS PAGE TO A FRIEND
Bioresource technology
Comparative studies on crosslinked and uncrosslinked natural rubber biodegradation by Pseudomonas sp.
PMID 16854583
Abstract
A comparative study on biodegradation of di-cumyl peroxide (DCP) crosslinked and uncrosslinked natural rubber by Pseudomonas sp. was carried out. Decrease in organic carbon content along with the changes in tensile strength of the treated rubber, both DCP crosslinked and uncrosslinked natural rubber, indicated rubber hydrocarbon utilization by the Pseudomonas sp. A decrease in 60.88% MPa and 41.66% MPa was observed after five month's old treated uncrosslinked natural rubber and DCP crosslinked rubber, respectively. Biodegradation was more pronounced in natural uncrosslinked rubber, which was further confirmed by the formation of aldehydic compounds with decrease in CH2 stretching frequencies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
329541
Dicumyl peroxide, 98%
C18H22O2