EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Design and synthesis of hydrophobic and chiral anions from amino acids as precursor for functional ionic liquids.
PMID 16855692
Abstract
Hydrophobic ionic liquids composed of tetrabutylphosphonium cation and chiral anions derived from amino acids modified with trifluoromethane sulfonyl groups have been synthesized using a simple method.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
189138
Tetrabutylphosphonium bromide, 98%
C16H36BrP
144800
Tetrabutylphosphonium chloride, 96%
C16H36ClP