EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Ruthenium-induced allylcarbamate cleavage in living cells.
PMID 16856188
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
736686
Allyl carbamate, 95%
C4H7NO2