EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Properties of a trifluoroleucine-resistant mutant of Saccharomyces cerevisiae.
PMID 16861814
Abstract
We characterized a trifluoroleucine-resistant mutant of Saccharomyces cerevisiae, TFL20, that has a mutation in the LEU4 gene. We monitored the concentration of extracellular i-AmOH and intracellular amino acids, and compared the ratios of gene expression in TFL20 with the wild-type strain, K30. We found that the LEU1, LEU2, and BAT1 genes were up-regulated in TFL20 for metabolism, and that TFL20 simultaneously produced as much i-AmOH and leucine as K30 does.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
91917
5,5,5-Trifluoro-DL-leucine, ≥98.0% (sum of isomers, HPLC)
C6H10F3NO2