EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Subcellular localization and possible functions of gamma-glutamyltransferase in the radish (Raphanus sativus L.) plant.
PMID 16861818
Abstract
Previously we reported the purification of soluble gamma-glutamyltransferases (GGTs) from radish cotyledon. Subcellular fractionation of radish cells revealed that soluble GGT is a vacuolar enzyme. Acivicin, a GGT inhibitor, mediated the in vivo catabolism inhibition of the glutathione S-conjugate generated from endogenous glutathione and exogenously supplied monochlorobimane. Thus soluble GGT is possibly involved in the catabolism of glutathione S-conjugates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
69899
Monochlorobimane, suitable for fluorescence, ≥70.0% (HPCE)
C10H11ClN2O2