EMAIL THIS PAGE TO A FRIEND
Oncogene
NF-kappaB represses E-cadherin expression and enhances epithelial to mesenchymal transition of mammary epithelial cells: potential involvement of ZEB-1 and ZEB-2.
PMID 16862183
Abstract
The transcription factor nuclear factor kappa B (NF-kappaB) is constitutively active in both cancer cells and stromal cells of breast cancer; however, the precise role of activated NF-kappaB in cancer progression is not known. Using parental MCF10A cells and a variant that expresses the myoepithelial marker p63 stably overexpressing the constitutively active p65 subunit of NF-kappaB (MCF10A/p65), we show that NF-kappaB suppresses the expression of epithelial specific genes E-cadherin and desmoplakin and induces the expression of the mesenchymal specific gene vimentin. P65 also suppressed the expression of p63 and the putative breast epithelial progenitor marker cytokeratin 5/6. MCF10A/p65 cells were phenotypically similar to cells undergoing epithelial to mesenchymal transition (EMT). MCF10A/p65 cells failed to form characteristic acini in three-dimensional Matrigel. Analysis of parental and MCF10A/p65 cells for genes previously shown to be involved in EMT revealed elevated expression of ZEB-1 and ZEB-2 in MCF10A/p65 cells compared to parental cells. In transient transfection assays, p65 increased ZEB-1 promoter activity. Furthermore, MCF10A cells overexpressing ZEB-1 showed reduced E-cadherin and p63 expression and displayed an EMT phenotype. The siRNA against ZEB-1 or ZEB-2 reduced the number of viable MCF10A/p65 but not parental cells, suggesting the dependence of MCF10A/p65 cells to ZEB-1 and ZEB-2 for cell cycle progression or survival. MCF10A cells chronically exposed to tumor necrosis factor alpha (TNFalpha), a potent NF-kappaB inducer, also exhibited the EMT-like phenotype and ZEB-1/ZEB-2 induction, both of which were reversed following TNFalpha withdrawal.