EMAIL THIS PAGE TO A FRIEND
Hellenic journal of cardiology : HJC = Hellenike kardiologike epitheorese
Targeting sarcoplasmic reticulum calcium handling proteins as therapy for cardiac disease.
PMID 16862819
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV51356
Anti-ASPH antibody produced in rabbit, affinity isolated antibody