EMAIL THIS PAGE TO A FRIEND
Human & experimental toxicology
Anti-clastogenic effect of beta-glucan extracted from barley towards chemically induced DNA damage in rodent cells.
PMID 16866189
Abstract
beta-Glucan (BG) was tested in vitro to determine its potential clastogenic and/or anti-clastogenic activity, and attempts were made to elucidate its possible mechanism of action by using combinations with an inhibitor of DNA polymerase. The study was carried out on cells deficient (CHO-k1) and cells proficient (HTC) in phases I and II enzymes, and the DNA damage was assessed by the chromosomal aberration assay. BG did not show a clastogenic effect, but was anti-clastogenic in both cell lines used, and at all concentrations tested (2.5, 5 and 10 microg/mL) in combination with damage inducing agents (methylmethane sulfonate in cell line CHO-k1, and methylmethane sulfonate or 2-aminoanthracene in cell line HTC). BG also showed a protective effect in the presence of a DNA polymerase beta inhibitor (cytosine arabinoside-3-phosphate, Ara-C), demonstrating that BG does not act through an anti-mutagenic mechanism of action involving DNA polymerase beta.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A38800
2-Aminoanthracene, 96%
C14H11N