EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacy and pharmacology
Influence of standard and nicotine-reduced cigarette smoke on plasma concentrations of isosorbide dinitrate and its metabolites in rats.
PMID 1686914
Abstract
Rats dosed orally with isosorbide dinitrate (1 mg kg-1) were exposed to smoke from standard and nicotine-reduced cigarettes for 8 min using a smoking machine. Plasma concentrations of isosorbide dinitrate and 5-isosorbide mononitrate, one of its major metabolites were approximately equal in the exposed groups, but were lower than in the non-smoking control group. The 2-isosorbide mononitrate concentration was also lower in the group exposed to smoke from standard cigarettes. Since the pharmacokinetics were influenced by smoke from both types of cigarette smoke, the effect may be attributed in large part to non-nicotine components of the smoke.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I0775020 Isosorbide 2-nitrate, European Pharmacopoeia (EP) Reference Standard
C6H9NO6