EMAIL THIS PAGE TO A FRIEND
Drug metabolism and disposition: the biological fate of chemicals
Identification and structure characterization of S-containing metabolites of 3-tert-butyl-4-hydroxyanisole in rat urine and liver microsomes.
PMID 1687007
Abstract
After ip administration of 3-tert-butyl-4-hydroxyanisole (3-BHA) to rats, two previously undocumented metabolites 2-tert-butyl-5-methylthiohydroquinone (TBHQ-5-SMe) and 2-tert-butyl-6-methylthiohydroquinone (TBHQ-6-SMe) were identified in the urine by comparison with the authentic samples by GC/MS. In addition to these metabolites, 3-tert-butyl-4,5-dihydroxyanisole was also detected in the urine hydrolyzed by beta-glucuronidase/sulfatase. Administration of tert-butylhydroquinone (TBHQ), an O-demethylated metabolite of 3-BHA, also resulted in the formation of the S-containing metabolites, TBHQ-5-SMe and TBHQ-6-SMe. After incubation of TBHQ with rat liver microsomes in the presence of glutathione (GSH), two metabolites were isolated and purified by HPLC. The metabolites were identified as 2-tert-butyl-5-(glutathion-S-yl)hydroquinone and 2-tert-butyl-6-(glutathion-S-yl)hydroquinone by 1H- and 13C-NMR spectrometry and by fast atom bombardment-mass spectrometry. The formation of TBHQ-GSH conjugates required NADPH, molecular oxygen, and GSH. Cytochrome P-450 inhibitors such as SKF 525-A and metyrapone markedly inhibited the formation of TBHQ-GSH conjugates in vitro. These results suggest that TBHQ is converted by cytochrome P-450-mediated monooxygenases to a reactive metabolite, 2-tert-butyl-p-benzoquinone (TBQ), which then conjugates with GSH to form TBHQ-GSH conjugates. GSH S-transferase activities do not seem to play a role in GSH conjugation reaction to TBQ because cytosol fraction from rat liver homogenates did not enhance the microsome-mediated production of TBHQ-GSH conjugates.