EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
The methionine salvage pathway compound 4-methylthio-2-oxobutanate causes apoptosis independent of down-regulation of ornithine decarboxylase.
PMID 16870157
Abstract
4-Methylthio-2-oxobutanoic acid (MTOB) is the final compound of the methionine salvage pathway that converts the polyamine byproduct methylthioadenosine to adenine and methionine. Here we find that MTOB inhibits growth of several human cell lines in a dose-dependent manner. Growth inhibition was specific for MTOB as we did not observe any inhibition with other chemically related compounds. MTOB treatment causes apoptosis and reduction of ornithine decarboxylase (ODC) activity but not ODC mRNA. To determine if MTOB exerts its effects primarily via ODC inhibition, we compared the effects of MTOB with the ODC-specific inhibitor difluoromethylornithine (DFMO). We found that MTOB was a more potent inducer of apoptosis than DFMO, lacked activation of caspase 3/7, and was able to induce apoptosis in cells lacking p53. Our results show that MTOB-induced growth inhibition and apoptosis is not simply secondary due to ODC inhibition and implies that MTOB activates apoptosis via other mechanisms.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K6000
α-Keto-γ-(methylthio)butyric acid sodium salt, ≥97%
C5H7NaO3S