EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry
Synthesis and antitumor activity of novel 10-substituted camptothecin analogues.
PMID 16870453
Abstract
In an attempt to improve the antitumor activity and decrease the cytotoxicity of camptothecin, 18 new 10-substituted camptothecin derivatives were prepared. The cytotoxicity in vitro on cancer cell lines and antitumor activity in vivo, and inhibitory properties of topoisomerase I of these derivatives were evaluated. Most of these derivatives possessed lower cytotoxicities than CPT, and the compounds 13, 21, 22, 23, and 24 showed similar topoisomerase I inhibitory activity to CPT. Analogues 13 exhibited the best antitumor activity in vivo among all derivatives we prepared.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
385344
4-Hydroxy-6-(trifluoromethyl)pyrimidine, 99%
C5H3F3N2O