EMAIL THIS PAGE TO A FRIEND
Therapie
Glycoprotein changes in non-insulin dependent diabetic rats: effect of N-benzoyl-D-phenylalanine and metformin.
PMID 16886705
Abstract
The effect of N-benzoyl-D-phenylalanine (NBDP) and metformin on neonatal streptozotocin (nSTZ) induced diabetes has been studied on plasma and tissue glycoproteins. In some pathological conditions, such as cancer, rheumatoid arthritis and diabetes, there is an abnormal glycosylation of acute phase serum proteins. As most serum proteins are produced in the liver, we have examined glycoprotein metabolism in diabetic condition. To induce non-insulin-dependent diabetes mellitus (NIDDM) a single dose of streptozotocin (100 mg/kg body weight) was injected into two day old rats. After 10-12 weeks, rats weighing above 150 g were selected for NIDDM model. In these rat, blood glucose and plasma glycoproteins were significantly increased whereas plasma insulin was significantly decreased. There was a significant decrease in the level of sialic acid and elevated levels of hexose, hexosamine and fucose in tissues. Oral administration of NBDP and metformin to diabetic rats decreased blood glucose and plasma glycoproteins. Plasma insulin and tissue sialic acid were increased whereas tissue concentrations of hexose, hexosamine and fucose were near normal. Our study suggests that NBDP and metformin possess a significant beneficial effect on glycoproteins in addition to their antidiabetic effect.