EMAIL THIS PAGE TO A FRIEND
Endocrinology
Differential expression of activator protein-1 proteins in the pineal gland of Syrian hamster and rat may explain species diversity in arylalkylamine N-acetyltransferase gene expression.
PMID 16887909