EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Isolation and mass spectrometry of specific DNA binding proteins.
PMID 16888366
Abstract
A subset of the proteome that binds to specific DNA sequences is at the center of genome function, integrity, and dynamics. We present a detailed protocol that allows the isolation of any specific DNA binding protein and its subsequent identification by mass spectrometry. The procedure involves prefractionation of crude nuclear extract by phosphocellulose (P11) chromatography, followed by a series of positive/negative selections on wild-type and site-mutated ligand DNA in a magnetic microparticulate format. DNA-affinity capture requires concatamerized and biotinylated ligand, selective salt conditions, and improved competitor DNAs. The amount of protein(s) captured on DNA-magnetic beads is generally sufficient for successful MALDI-TOF and TOF/TOF MS-based protein identification. As an example, we describe the procedures used to isolate and identify four specific transcription factors from 2 x 10(9) promyelocytic NB4 cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2258 Cellulose phosphate, (Fine Mesh)