EMAIL THIS PAGE TO A FRIEND
The Journal of endocrinology
Characterisation of the prereceptor regulation of glucocorticoids in the anterior segment of the rabbit eye.
PMID 16899581
Abstract
The prereceptor regulation of glucocorticoids (GCs) by 11beta-hydroxysteroid dehydrogenase type-1 (11beta-HSD1), a bidirectional isozyme that interconverts active (cortisol) and inactive (cortisone) GCs, is an established determinant of GC function in tissues such as liver, adipose and bone. Although the therapeutic use of GCs is abundant in ophthalmic practice, where GC interactions with nuclear receptors modulate gene transcription, the prereceptor regulation of endogenous cortisol is not well described in ocular tissues. Recent descriptive studies have localised 11beta-HSD1 to the human corneal epithelium and non-pigmented epithelium (NPE) of the ciliary body, indicating a link to corneal epithelial physiology and aqueous humour production. In this study, we characterise the functional aspects of the autocrine regulation of GCs in the anterior segment of the rabbit eye. Using our in-house generated primary antibody to human 11beta-HSD1, immunohistochemical analyses were performed on paraffin-embedded sections of whole New Zealand white albino rabbits, (NZWAR) eyes. As in human studies, 11beta-HSD1 was localised to the corneal epithelium and the NPE. No staining was seen in the albino 'pigmented' ciliary epithelium. Specific enzyme assays for oxo-reductase (cortisone-->cortisol) and dehydrogenase (cortisol-->cortisone) activity indicated predominant 11beta-HSD1 oxo-reductase activity from both the intact ciliary body tissue (n=12, median 2.1 pmol/mg per h and range 1.25-2.8 pmol/mg per h; P=0.006) and primary cultures of corneal epithelial cells (n=12, median 3.0 pmol/mg per h and range 1.0-7.4 pmol/mg per h, P=0.008) compared with dehydrogenase activity (median 1.0 pmol/mg per h and range 0.5-2.0 pmol/mg per h; median 0.5 pmol/mg per h and range 0.25-1.9 pmol/mg per h respectively). These findings were supported by expression of 11beta-HSD1 protein as visualised by Western blotting of ciliary body tissue and immunocytochemistry of corneal epithelial cells. Reduction of corneal epithelial cell proliferation was seen after primary cultures were co-incubated with cortisol and cortisone. 11beta-HSD1 activity was not demonstrated in naïve conjunctival fibroblasts or corneal stromal keratocytes. Our results indicate that the distribution of 11beta-HSD1 in the rabbit resembles that of the human eye and activates cortisone to cortisol in both corneal and uveal tissues. The NZWAR provides a suitable in vivo model for the further evaluation of 11beta-HSD1 activity in the eye, especially its role in corneal epithelial and ciliary body physiology.