EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Substituted 5-benzyl-2-phenyl-5H-imidazo[4,5-c]pyridines: a new class of pestivirus inhibitors.
PMID 16901692
Abstract
A novel class of inhibitors of pestiviruses (5-substituted 2-phenyl-5H-imidazo[4,5-c]pyridines) is described. Modification of the substituent in position 5 resulted in analogues with high activity (EC(50)<100nM) and selectivity (SI>1000) against the pestivirus BVDV (bovine viral diarrhea virus).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
264431
2,6-Difluorobenzyl bromide, 97%
C7H5BrF2