EMAIL THIS PAGE TO A FRIEND
Circulation
Images in cardiovascular medicine. A rare form of midventricular Tako-Tsubo after emotional stress followed up with magnetic resonance imaging.
PMID 16908776
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T89605
Tropinone, 99%
C8H13NO