EMAIL THIS PAGE TO A FRIEND
Oncogene
Farnesyltransferase and geranylgeranyltransferase I inhibitors upregulate RhoB expression by HDAC1 dissociation, HAT association and histone acetylation of the RhoB promoter
PMID 16909123