EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[A synthesis of cinnamoyloxyisoflavones].
PMID 16909870
Abstract
Interaction of 7-hydroxyisoflavonones with cinnamoyl chloride results in cinnamoyloxyisoflavonones.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C81101
Cinnamoyl chloride, 98%
C9H7ClO