EMAIL THIS PAGE TO A FRIEND
Molecular therapy : the journal of the American Society of Gene Therapy
Parkin protects against neurotoxicity in the 6-hydroxydopamine rat model for Parkinson's disease.
PMID 16914382
Abstract
Loss-of-function mutations in the PARK2 gene are the major cause of early onset familial Parkinson's disease. The gene product, parkin, is an E3 ligase of the ubiquitin-proteasome pathway involved in protein degradation. Dopaminergic neuron loss may result from the toxic accumulation of parkin substrates, suggesting a key role for parkin in dopaminergic neuron survival. In this study, we have investigated the neuroprotective capacity of parkin in the 6-OHDA rat model for Parkinson's disease. 6-OHDA induces the generation of reactive oxygen species leading to the degeneration of catecholaminergic neurons, but may also impair proteasome activity. Lentiviral vectors encoding human wild-type parkin or green fluorescent protein were stereotactically injected into the substantia nigra 2 weeks prior to a striatal 6-OHDA lesion. Histological analysis 1 and 3 weeks after lesioning showed a significant preservation of dopaminergic cell bodies and nerve terminals. Moreover, lesioned rats overexpressing parkin displayed a corresponding behavioral improvement as measured by the amphetamine-induced rotation test and the cylinder test. The improved performance in the amphetamine-induced rotation test lasted until 20 weeks after lesioning. Our results demonstrate that parkin acts as a potent neuroprotective agent in vivo against 6-OHDA toxic insults. These data support the therapeutic potential of parkin for the treatment of not only familial but also sporadic Parkinson's disease.