EMAIL THIS PAGE TO A FRIEND
British journal of hospital medicine (London, England : 2005)
The evolving role of exemestane in the management of breast cancer.
PMID 16918105
Abstract
The role of therapies in the management of breast cancer is continuously changing. As research and development furthers, newer and more effective treatments are beginning to take the lead in cancer care, changing the role of tamoxifen.