EMAIL THIS PAGE TO A FRIEND
Langmuir : the ACS journal of surfaces and colloids
Micellization and drug solubilization in aqueous solutions of a diblock copolymer of ethylene oxide and phenyl glycidyl ether.
PMID 16922522
Abstract
The aim of this study was to define a block copolymer micellar system with a high solubilization capacity for poorly soluble aromatic drugs. Ethylene oxide and phenyl glycidyl ether were sequentially polymerized to form the diblock copolymer G5E67 (G = phenyl glycidyl ether, OCH2CH(CH2OC6H5); E = oxyethylene, OCH2CH2; subscripts denote number-average block lengths in repeat units). The association properties in aqueous solution over the range 20-50 degrees C were investigated by surface tensiometry and light scattering, yielding values of the cmc, hydrodynamic radius, and association number; gel boundaries in concentrated micellar solution were investigated by tube inversion. The solubilization capacity of G5E67 for the poorly water-soluble drug griseofulvin was higher than that of a triblock EGE copolymer of longer G block length and considerably higher than that achieved with poloxamers (EmPnEm, P = oxypropylene).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
248487
1,2-Epoxy-3-phenoxypropane, 99%
C9H10O2
578576
1,2-Epoxy-3-phenoxypropane, technical grade
C9H10O2