EMAIL THIS PAGE TO A FRIEND
Klinicheskaia meditsina
[The influence of the bioflanoid dicvertin on the antioxidative system ceruloplasmin-transferrin and lipid peroxidation in patients suffering from stable coronary heart disease with dyslipidemia].
PMID 16924801
Abstract
Thirty-two patients with coronary heart disease (CHD)--exertional angina and postinfarction cardiosclerosis with dislipidemia--were treated with dicvertin (2,3-dihydro-3,5,7-trihydroxi-2(3,4-dihydroxiphenyl)-4H-1-benzopyran-4-on), a Russian antioxidant, during two months in a day dose of 80 mg in addition to conventional cardial therapy. The therapy resulted in positive changes including improvement of CHD clinical picture and biochemical serum indices, such as a 6% decrease in cholesterol level, a 12% decrease in low-density lipoproteins cholesterol level, and a 14% increase in high-density lipoproteins cholesterol level. The intensity of lipid peroxidation decreased, the levels of diene conjugates and TBA-reactive products lowered by 38% and 40%, respectively. The fibrinogen level lowered by 20%. The antioxidative status of the patients increased.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68530
Pinobanksin, ≥95% (HPLC)
C15H12O5