EMAIL THIS PAGE TO A FRIEND
Yao xue xue bao = Acta pharmaceutica Sinica
[Analysis of anisodine and its metabolites in rat plasma by liquid chromatography-tandem mass spectrometry].
PMID 16927825
Abstract
To identify anisodine and its metabolites in rat plasma after ingestion of anisodine by combining liquid chromatography and tandem mass spectrometry (LC-MS(n)). Plasma samples from rats after a single orally administration of 20 mg anisodine were added with methanol to precipitate protein. Then, it was analyzed by LC-MS(n). Identification and structural elucidation of the metabolites were performed by comparing their changes in molecular masses, retention-times and full scan MS(n) spectra with those of the parent drug and blank plasma. The results revealed that the parent drug and its four metabolites (norscopine, scopine, hydroxyanisodine, N-oxide anisodine) existed in rat plasma. This method is sensitive, rapid, simple, and it is suitable for the rapid identification of drug and its metabolits.