EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Microwave-assisted synthesis of thieno[2,3-c]pyridine derivatives as a new series of allosteric enhancers at the adenosine A(1) receptor.
PMID 16931014
Abstract
The microwave-assisted aromatization method has been used for the synthesis of a series of novel thieno[2,3-c]pyridines. This rapid method produces compounds in good yield within minutes in comparison with conventional heating method. The synthesized molecules have been evaluated as a potential new series of allosteric enhancers acting at the adenosine A(1) receptor. In a functional assay, one compound (3h) showed activity comparable with that of reference compound PD 81,723.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1123
PD 81,723, ≥98% (HPLC), solid
C14H12F3NOS