EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Oxidative DNA and RNA damage in rat liver due to acetoxime: similarity to effects of 2-nitropropane.
PMID 1693319
Abstract
Acetoxime (ACO) and 2-nitropropane (2-NP), both industrially important chemicals and known hepatocarcinogens in rats, induced increased levels of 8-hydroxy-guanine in liver DNA and RNA of male Sprague-Dawley and F344 rats after either oral or i.p. administration. Both compounds also produced qualitatively the same patterns of other apparent modifications of liver DNA and RNA nucleosides, discernible by HPLC with electrochemical detection. Six hours after administration, the effects of 2-NP on liver nucleic acids were more pronounced in F344 rats than in Sprague-Dawley rats, suggesting that 2-NP may prove to be a stronger carcinogen in the F344 strain. The effects of ACO, a weaker carcinogen than 2-NP, were less than those of the nitroalkane in both rat strains. These results suggest that the hepatocarcinogenicity of ACO, like that of 2-NP, may depend on increased generation of reactive oxygen species capable of producing DNA and RNA base damage in rat liver. In addition, the data support the hypothesis that the hepatocarcinogenicity of ACO depends on its partial in vivo N-oxidation to 2-NP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00750
Acetone oxime, purum, ≥98.0% (GC)
C3H7NO
A10507
Acetone oxime, 98%
C3H7NO