EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Epibatidine isomers and analogues: structure-activity relationships.
PMID 16934977
Abstract
Binding affinities for a range of epibatidine isomers and analogues at the alpha4beta2 and alpha3beta4 nAChR subtypes are reported; compounds having similar N-N distances to epibatidine show similar, high potencies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C2899
Cytisine, ≥99%, powder
C11H14N2O