EMAIL THIS PAGE TO A FRIEND
Chemico-biological interactions
Effect of n-hexane on the disposition and toxicity of the 1,3-butadiene metabolite 3-butene-1,2-diol.
PMID 16935275
Abstract
3-Butene-1,2-diol (butenediol), a major metabolite of 1,3-butadiene (butadiene), can undergo either detoxification or biotransformation to potentially toxic metabolites, including 3,4-epoxy-1,2-butanediol and hydroxymethylvinyl ketone (HMVK). Butadiene exposure can occur concomitantly with hexanes, which share common biotransformation pathways with butadiene. To determine the potential influence of hexane co-exposure on butadiene toxicity, the present study examined the effect of n-hexane on butenediol disposition [as measured by urinary excretion of (N-acetyl-S-(3,4-dihydroxybutyl)-L-cysteine) (MI level)] and genotoxicity (as measured by the frequency of bone marrow micronucleated erythrocytes) and acute toxicity (as measured by body weight changes) in the rat. The results show that butenediol was not genotoxic to adult or immature rats but was acutely toxic to adult but not immature rats. The results also suggest that n-hexane co-exposure may attenuate the acute toxicity by butenediol in adult rats and that immature rats may be less sensitive than adults to the acute toxicity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
488216
3,4-Dihydroxy-1-butene, ≥99%
C4H8O2