EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Fractionation of whale cartilage chondroitin sulfate on Sepharose CL-4B in the presence of high concentration of ammonium sulfate. Relationship between molecular size and unit-disaccharide composition of the polysaccharide.
PMID 1693882
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CL4B200 Sepharose® CL-4B, Cross-linked