EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
High-precision catalysts: regioselective hydroformylation of internal alkenes by encapsulated rhodium complexes.
PMID 16939244
Abstract
We report the formation of high-precision catalysts using encapsulated rhodium complexes. In the current example, the encapsulated rhodium catalyst shows unprecedented high selectivity in the rhodium-catalyzed hydroformylation of internal alkenes, forming predominantly one of the branched aldehydes. This catalyst system is the first example that is able to discriminate between carbon atoms C3 and C4 in trans-3-octene.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74900
1-Octene, analytical standard
C8H16
O4806
1-Octene, 98%
C8H16
W429300
1-Octene, 98%
C8H16