EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Redefining the structure-activity relationships of 2,6-methano-3-benzazocines. 4. Opioid receptor binding properties of 8-[N-(4'-phenyl)-phenethyl)carboxamido] analogues of cyclazocine and ethylketocycalzocine.
PMID 16942039
Abstract
The synthesis and evaluation of a series of aryl-containing N-monosubstituted analogues of the lead compound 8-carboxamidocyclazocine were performed to probe a putative hydrophobic binding pocket of opioid receptors. High binding affinity to mu, kappa, and delta opioid receptors was observed for the 8-[N-(4'-phenyl)-phenethyl)carboxamido] analogue.