EMAIL THIS PAGE TO A FRIEND
Bioorganic chemistry
Synthesis of pyrrolo[2,3-d]pyrimidine nucleoside derivatives as potential anti-HCV agents.
PMID 16945403
Abstract
Several Toyocamycin (4) analogues were examined for their ability to inhibit HCV RNA in a replicon assay. Among the compounds examined 4-methylthio (18) and 5-carboxamide oxime derivatives (23 and 27) of Toyocamycin were found to have good activity and selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3580
Toyocamycin, ≥98% (HPLC), from Streptomyces rimosus
C12H13N5O4