EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Inhibitory effect of ammonium tetrathiotungstate on tyrosinase and its kinetic mechanism.
PMID 16946532
Abstract
Tyrosinase requires two copper ions at the active site, in order to oxidize phenols to catechols. In this study, the inhibitory effect of the copper-chelating compound, ammonium tetrathiotungstate (ATTT), on the tyrosinase activity was investigated. ATTT was determined to inactivate the activity of mushroom tyrosinase, in a dose-dependent manner. The kinetic substrate reaction revealed that ATTT functions as a kinetically competitive inhibitor in vitro, and that the enzyme-ATTT complex subsequently undergoes a reversible conformational change, resulting in the inactivation of tyrosinase. In human melanin-producing cells, ATTT evidenced a more profound tyrosinase-inhibitory effect than has been seen in the previously identified tyrosinase inhibitors, including kojic acid and hydroquinone. Our results may provide useful information for the development of whitening agent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
336734
Ammonium tetrathiotungstate, ≥99.9% trace metals basis
H8N2S4W