EMAIL THIS PAGE TO A FRIEND
Cancer research
Fluorodeoxyuridine modulates cellular expression of the DNA base excision repair enzyme uracil-DNA glycosylase.
PMID 16951200
Abstract
The thymidylate synthase inhibitor 5-fluorouracil (5-FU) continues to play a pivotal role in the treatment of cancer. A downstream event of thymidylate synthase inhibition involves the induction of a self-defeating base excision repair process. With the depletion of TTP pools, there is also an increase in dUMP. Metabolism of dUMP to the triphosphate dUTP results in elevated pools of this atypical precursor for DNA synthesis. Under these conditions, there is a destructive cycle of dUMP incorporation into DNA, removal of uracil by the base excision repair enzyme uracil-DNA glycosylase (UDG), and reincorporation of dUMP during the synthesis phase of DNA repair. The end point is DNA strand breaks and loss of DNA integrity, which contributes to cell death. Evidence presented here indicates that both the nuclear and the mitochondrial isoforms of UDG are modulated by FdUrd (and 5-FU) treatment in certain cell lines but not in others. Modulation occurs at the transcriptional and post-translational levels. Under normal conditions, nUDG protein appears in G(1) and is degraded during the S to G(2) phase transition. The present study provides evidence that, in certain cell lines, FdUrd mediates an atypical turnover of nUDG. Additional data indicate that, for cell lines that do not down-regulate nUDG, small interfering RNA-mediated knockdown of nUDG significantly increases resistance to the cytotoxic effects of FdUrd. Results from these studies show that nUDG is an additional determinant in FdUrd-mediated cytotoxicity and bolster the notion that the self-defeating base excision repair pathway, instigated by elevated dUTP (FdUTP) pools, contributes to the cytotoxic consequences of 5-FU chemotherapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3876
2′-Deoxyuridine 5′-monophosphate disodium salt, Sigma Grade
C9H11N2O8P · 2Na