EMAIL THIS PAGE TO A FRIEND
Journal of environmental monitoring : JEM
Aquatic processes and systems in perspective.
PMID 16951746
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
60720
Ketone Moschus, ≥97.0% (N)
C14H18N2O5
W522902
Musk ketone, ≥98%
C14H18N2O5