EMAIL THIS PAGE TO A FRIEND
Agents and actions
A modified radioimmunoassay for the detection of histamine release in vivo and in vitro in man.
PMID 1695451
Abstract
Histamine (Hi) release in vitro or in vivo in man was measured by variants of a radioimmunoassay (RIA) procedure. Hi released from isolated basophils was converted enzymically to N-tau-methylhistamine (NMH) which was then measured by a very sensitive RIA. This modified RIA was compared with the standard spectrofluorometric assay and was found to have additional advantages in certain applications. RIA of NMH in plasma was found to be of value in acute medical conditions of obscure aetiology.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7780
3-Methylhistamine dihydrochloride, analytical standard
C6H11N3 · 2HCl