EMAIL THIS PAGE TO A FRIEND
Current opinion in investigational drugs (London, England : 2000)
Glycopeptides and glycodepsipeptides in clinical development: a comparative review of their antibacterial spectrum, pharmacokinetics and clinical efficacy.
PMID 16955686
Abstract
Hemi-synthetic derivatives of glycopeptides have demonstrated bactericidal activity towards Gram-positive bacteria, including vancomycin-resistant strains (oritavancin and telavancin), and a prolonged half-life, allowing for once-daily (oritavancin and telavancin) or once-weekly (dalbavancin) administration. These compounds have proved effective for the treatment of infections caused by multidrug-resistant Gram-positive bacteria, including complicated skin and skin structure infections (oritavancin, telavancin and dalbavancin), bacteremia (oritavancin and dalbavancin) and nosocomial pneumonia. This review compares the antibacterial activity and clinical activity of three glycopeptides, oritavancin, telavancin and dalbavancin, and the natural lipoglycopeptide, ramoplanin, which, being unstable in the bloodstream, is administered orally to treat Clostridium difficile colitis and for digestive tract decontamination. All of these compounds, with the exception of oritavancin, have received Fast Track designation from the FDA because of their clinical efficacy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R1781 Ramoplanin