EMAIL THIS PAGE TO A FRIEND
Biomacromolecules
Self-assembly of monoglycerides in beta-lactoglobulin adsorbed films at the air-water interface. Structural, topographical, and rheological consequences.
PMID 16961330
Abstract
In this work, we have analyzed the structural, topographical, and surface dilatational characteristics of pure beta-lactoglobulin adsorbed films and the effect of the self-assembly of monoglycerides (monopalmitin or monoolein) in beta-lactoglobulin films at the air-water interface. Measurements were performed in a single device that incorporates a Wilhelmy-type film balance, Brewster angle microscopy, and interfacial dilatational rheology. The structural and topographical characteristics of beta-lactoglobulin adsorbed and spread films are similar. However, the surface dilatational modulus of beta-lactoglobulin films shows a complex behavior depending on film formation. The self-assembly of monoglyceride in a beta-lactoglobulin adsorbed film has an effect on the structural, topographical, and dilatational properties of the mixed films, depending on the interfacial composition and the surface pressure (pi). At low pi, a mixed film of monoglyceride and beta-lactoglobulin may exist. At high pi (after the collapse of beta-lactoglobulin), the mixed films are dominated by monoglyceride molecules. However, the small amounts of collapsed beta-lactoglobulin have a significant effect on the surface dilatational properties of the mixed films. Protein displacement by monoglyceride is higher for monopalmitin than for monoolein. However, some degree of interaction exists between proteins and monoglycerides, and these interactions are more evident in adsorbed films than in spread films.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M1640
DL-α-Palmitin, ≥99%
C19H38O4