EMAIL THIS PAGE TO A FRIEND
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology
Silica-guanidinium thiocyanate-based nucleic acid isolation protocol does not improve sensitivity of two commercial tests for detection of Mycobacterium tuberculosis in respiratory samples.
PMID 16964511