EMAIL THIS PAGE TO A FRIEND
Cell Biology and Toxicology
Expression and transcriptional activities of nuclear receptors involved in regulation of drug-metabolizing enzymes are not altered by colchicine: Focus on PXR, CAR, and GR in primary human hepatocytes
PMID 16964586
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AB10303 Anti-CAR Antibody