EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Selective monoiodination of aromatic compounds with electrochemically generated I+ using micromixing.
PMID 16969460
Abstract
Selective monoiodination of aromatic compounds such as dimethoxybenzene has been successfully achieved with I+, which is generated by anodic oxidation of I2 in acetonitrile, using micromixing.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W238600
1,4-Dimethoxybenzene, 99%, FG
C8H10O2
D131350
1,4-Dimethoxybenzene, ReagentPlus®, 99%
C8H10O2