EMAIL THIS PAGE TO A FRIEND
Advance for nurse practitioners
Once-daily intranasal corticosteroids for allergic rhinitis. Examining Treatment issues.
PMID 16972477
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5021
Flunisolide, ≥97%
C24H31FO6