EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Aryl C-glycosylation of phenols with glycosyl trifluoroacetimidates.
PMID 16978594
Abstract
Aryl C-glycosylation of a variety of phenols with glycosyl trifluoroacetimidates in the presence of TMSOTf was examined, leading to the corresponding ortho-hydroxyaryl C-glycosides in variable yields.