EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Structural organization of [met]enkephalin and endorphin molecules. II. Theoretical conformation analysis of alpha-, gamma- and delta-endorphins].
PMID 1698060
Abstract
Conformational energy calculations were carried out for neuropeptides alpha-, gamma- and delta-endorphins, which are 1-16, 1-17 and 1-19 fragments respectively, of beta-endorphin. The proposed computational scheme yielded all possible low-energy conformational sets for these hormones. Specific features of spatial organization of each compound and similarities of their structures are discussed.