EMAIL THIS PAGE TO A FRIEND
Lipids
Single-step analysis of individual conjugated bile acids in human bile using 1H NMR spectroscopy.
PMID 16981437
Abstract
1H and 13C NMR spectra of intact human bile were assigned using one-dimensional (1H and 13C) and two-dimensional (1H-1H and 1H-13C) experiments. Individual conjugated bile acids--glycocholic acid, glycodeoxycholic acid, glycochenodeoxycholic acid, taurocholic acid, taurodeoxycholic acid, and taurochenodeoxycholic acid--were identified. The bile acids were quantified accurately and individually in a single step by using distinct and characteristic amide signals. Making use of 13C NMR, the study also suggests a way to analyze unconjugated bile acids separately, if present. Chemical shift assignments and rapid single-step analysis of individual conjugated bile acids from intact bile presented herein may have immense utility in the study of bile acid metabolism and deeper understanding of hepatobiliary diseases.