EMAIL THIS PAGE TO A FRIEND
Bibliotheca haematologica
Suppression of production of Epstein-Barr virus in P3HR-1 cell line by bovine serum.
PMID 169827
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
P3HR-1, 86010701